Dithiothreitol (DTT, Cleland's reagent) 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Warning

32719-0100 32719-0500 32719-0010
TS32719-0100EA 375 USD
TS32719-0100 TS32719-0500 TS32719-0010
Dithiothreitol (DTT, Cleland's reagent) 99+% for molecular biology DNase-, RNase-, protease-free
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
Boiling Pt: 125…130 °C (2 mmHg)
Melting Pt: 39…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
UN: 3335
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Titration Iodometric 99.0 to 101.0 %
UV (0.1 M in water)
Dnase,Rnase,Protease activity none detected
Heavy metals (as Pb) ≤5 ppm
Oxidized DTT(UV) ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR