N,N-Dimethylformamide 99.8% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Danger

327170025 327170100 327171000 327175000
TS327170025EA 232 USD
TS327170025 TS327170100 TS327171000 TS327175000
N,N-Dimethylformamide 99.8% for molecular biology DNase-, RNase-, protease-free
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.945 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.8 %
UV at 280 nm A: ≤0.1
Color scale ≤5 APHA
Residue after evaporation ≤0.001 %
Dnase,Rnase,Protease activity none detected
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Free amine ≤0.001 % (Kaiser)
Acidity (CH3COOH) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR