Phenol liquefied/tris saturated solution stabilized for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Danger

32712-5000
TS32712-5000EA 266 USD
TS32712-5000
Phenol liquefied/tris saturated solution stabilized for molecular biology DNase-, RNase-, protease-free
Phenol liquefied/Tris
Formula: C₆H₅OH
Storage Temperature: Refrigerator
MDL Number: MFCD01632719
CAS Number: 108-95-2
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow to orange Solution
Water 25 to 35 % (K.F.)
Stabilizer 0.08 to 0.12 % hydroxyquinoline
pH 7.8 to 8.2 (at 25°C)
Dnase,Rnase,Protease activity None detected
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR