Methanol, extra dry 99.9%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

32695-0010 32695-0025 32695-1000
TS32695-0010EA 96 USD
TS32695-0010 TS32695-0025 TS32695-1000
Methanol, extra dry 99.9%, AcroSeal™
Methanol
Formula: CH₃OH
Boiling Pt: 64.7 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.85 %
Residue after evaporation ≤0.0005 %
Acidity/alkalinity ≤0.0005 meq/g
Filt.over 0.2 micron PFTE Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR