Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

325410100
TS325410100EA 182 USD
TS325410100
Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy
Acetonitrile-[D3]

Packaged in 1.00 ml ampoules

Formula: D₃CCN
Boiling Pt: 80.7 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR