2-(N-Ethylanilino)ethanol 96%

Supplier: Thermo Scientific Chemicals
Warning

32522-1000
TS32522-1000EA 111 USD
TS32522-1000
2-(N-Ethylanilino)ethanol 96%
2-(N-Ethylanilino)ethanol
Formula: C₁₀H₁₅NO
Boiling Pt: 276 °C
Melting Pt: 33 °C
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00020575
CAS Number: 92-50-2
EINECS: 202-160-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid or solid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5640 (20 °C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR