Dimethyl sulfoxide-[D₆] 100.0 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulfoxide, Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulfoxide-[²H₆], Dimethyl sulphoxide-d6, Dimethyl sulfoxide-d6

320760075
TS320760075EA 203 USD
TS320760075
Dimethyl sulfoxide-[D₆] 100.0 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]

Packaged in 0.75 ml ampoules

Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³
Flash Pt: 88 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR