N,N-Dimethylformamide-[D₇] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7

32073-0075
TS32073-0075EA 407 USD
TS32073-0075
N,N-Dimethylformamide-[D₇] 99.5 atom % D for NMR spectroscopy
N,N-Dimethylformamide-[D7]
Formula: DCOON(CD₃)₂
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.03 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR