Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

32072-0075
TS32072-0075EA 276 USD
TS32072-0075
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D2]
Formula: CD₂Cl₂
Boiling Pt: 40 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR