Deuterium oxide 100.0 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Water-D2, Heavy water

32070-0075
TS32070-0075EA 61 USD
TS32070-0075
Deuterium oxide 100.0 atom % D for NMR spectroscopy
Deuterium oxide

Packaged in 0.75 ml ampoules

Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.95 %
Tritium (T) ≤74 Bq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR