Chloroform-[D₁] 100.0 atom % D NMR

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

320680075
TS320680075EA 78 USD
TS320680075
Chloroform-[D₁] 100.0 atom % D NMR
Chloroform-[D1]

Packaged in 0.75 ml ampoules

Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8
Melting Pt: -64.1
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.01 % (Coulometric)
D-Enrichment ≥99.95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR