Acetone-[D₆] 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

320630075
TS320630075EA 43 USD
TS320630075
Acetone-[D₆] 99.8 atom % D for NMR spectroscopy
Acetone-[D6]

Packaged in 0.75 ml ampoules

Formula: D₃CC(O)CD₃
Boiling Pt: 55.5 °C (1013 hPa)
Melting Pt: –93 °C
Density: 0.87 g/cm³
Flash Pt: –20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR