L(-)-Glutathione (oxidised form) 98%

Supplier: Thermo Scientific Chemicals

32022-0050 32022-5000
TS32022-0050EA 252 USD
TS32022-0050 TS32022-5000
L(-)-Glutathione (oxidised form) 98%
L(-)-Glutathione (oxidized form)
Formula: C₂₀H₃₂N₆O₁₂S₂
Melting Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8
EINECS: 248-170-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤8 % (105°C, 3 h)
HPLC ≥97.5 %
Residue after ignition ≤0.5 %
Specific optical rotation -92° to -106° (20°C, 589 nm) (c=4, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR