Tris(dibenzylideneacetone)dipalladium(0) 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]

31877-0050 AC31877-0250 AC31877-5000
TS31877-0050EA 246 USD
TS31877-0050 TS31877-0250 TS31877-5000
Tris(dibenzylideneacetone)dipalladium(0) 97%
Tris(dibenzylideneacetone)dipalladium(0)
Formula: C₅₁H₄₂O₃Pd₂
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00013310
CAS Number: 51364-51-3

Order Now

Specification Test Results

Appearance Purple to black Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR