Gallium 99.99% (trace metals basis)
Supplier: Thermo Scientific Chemicals
31799-0500
317990100
TS31799-0500EA
285
USD
TS31799-0500
TS317990100
Gallium 99.99% (trace metals basis)
Gallium
Specification Test Results
Appearance | Silver white to silver grey Metallic solid or liquid |
Total trace metal impurities | ≤0.01 % |