Silver 99.9% (trace metals basis), wire, Diameter 0.25 mm , 1ea = 10.4gr

Supplier: Thermo Scientific Chemicals

317730010
TS317730010EA 179 USD
TS317730010
Silver 99.9% (trace metals basis), wire, Diameter 0.25 mm , 1ea = 10.4gr
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961 °C
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Appearance Silver white Wire
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR