Silver 99.9% Trace metals basis, wire, Diameter 2.0 mm , 1ea = 16.5gr

Supplier: Thermo Scientific Chemicals

317720010
TS317720010EA 188 USD
TS317720010
Silver 99.9% Trace metals basis, wire, Diameter 2.0 mm , 1ea = 16.5gr
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Appearance Silver white Wire
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR