Manganese 99+%, powder -40 mesh

Supplier: Thermo Scientific Chemicals
Warning

317440010 317442500
TS317440010EA 105 USD
TS317440010 TS317442500
Manganese 99+%, powder -40 mesh
Manganese
Formula: Mn
Boiling Pt: 1900 °C
Melting Pt: 1260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,05762

Order Now

Specification Test Results

Appearance Grey-brown to brown-black Powder
Assay ≥99.0 % (Mn)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR