Manganese 99+%, powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
317440010
317442500
TS317440010EA
105
USD
TS317440010
TS317442500
Manganese 99+%, powder -40 mesh
Manganese
Formula:
Mn Boiling Pt: 1900 °C Melting Pt: 1260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.1,III Merck Index: 12,05762 |
Specification Test Results
Appearance | Grey-brown to brown-black Powder |
Assay | ≥99.0 % (Mn) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...