Triammonium dodecamolybdophosphate hydrate

Supplier: Thermo Scientific Chemicals
Warning

AC31710-0010 AC31710-0500 AC31710-2500
TS31710-0010EA 1156 USD
TS31710-0010 TS31710-0500 TS31710-2500
Triammonium dodecamolybdophosphate hydrate
Triammonium dodecamolybdophosphate hydrate
Formula: H₁₂Mo₁₂N₃O₄₀P·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150423
CAS Number: 54723-94-3
EINECS: 234-714-0

Order Now

Specification Test Results

Appearance Yellow Powder
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤250 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR