Iron(II) sulfide 99.9% (trace metals basis) -100 mesh
Supplier: Thermo Scientific Chemicals
Synonyms:
Ferrous sulphide, Ferrous sulfide
AC31587-0100
AC31587-0500
TS31587-0100EA
79
USD
TS31587-0100
TS31587-0500
Iron(II) sulfide 99.9% (trace metals basis) -100 mesh
Iron (II) sulphide
Formula:
FeS Melting Pt: 1195 °C Density: 4.84 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011013 CAS Number: 1317-37-9 EINECS: 215-268-6 Merck Index: 12,04106 |
Specification Test Results
Appearance | Gray to brown to black Powder |
Total trace metal impurities | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...