Magnesium hydroxide 95%, pure

Supplier: Thermo Scientific Chemicals

AC31572-0025 AC31572-0250 AC31572-5000
TS31572-0025EA 232 USD
TS31572-0025 TS31572-0250 TS31572-5000
Magnesium hydroxide 95%, pure
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Melting Pt: 350 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR