Copper(II) chloride dihydrate 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

AC31528-0025 AC31528-1000 AC31528-5000
TS31528-0025EA 927 USD
TS31528-0025 TS31528-1000 TS31528-5000
Copper(II) chloride dihydrate 99+% ACS
Copper (II) chloride dihydrate
Formula: CuCl₂·2H₂O
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Appearance Blue-green to blue Crystalline powder or crystals
Titration Iodometric ≥99.0 %
Nitrate (NO3) ≤150 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤50 ppm
Nickel (Ni) ≤100 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR