Chromium(II) chloride 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chromium dichloride
AC31519-0010
AC31519-0050
AC31519-0250
TS31519-0010EA
66
USD
TS31519-0010
TS31519-0050
TS31519-0250
Chromium(II) chloride 99.9% (trace metals basis)
Chromium (II) chloride
Formula:
Cl₂Cr Melting Pt: 824 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010947 CAS Number: 10049-05-5 EINECS: 233-163-3 Merck Index: 13,02264 |
Specification Test Results
Appearance | Off-white to gray-green Powder |
Total trace metal impurities | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...