Strontium nitrate 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

AC31517-0025 AC31517-1000 AC31517-5000
TS31517-0025EA 472 USD
TS31517-0025 TS31517-1000 TS31517-5000
Strontium nitrate 99+% ACS
Strontium nitrate
Formula: Sr(NO₃)₂
Melting Pt: 570 °C
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 13,08926

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric ≥99.0 %
Loss on drying ≤0.1 % (105°C)
pH 5.0 to 7.0 (5 % solution at 25.0°C)
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Barium (Ba) ≤500 ppm
Calcium (Ca) ≤500 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤1000 ppm
Sodium (Na) ≤1000 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR