1-Adamantyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

31436-0010 31436-0050 31436-0250
TS31436-0010EA 36 USD
TS31436-0010 TS31436-0050 TS31436-0250
1-Adamantyl isocyanate 98%
1-Adamantyl isocyanate

Appearance: White Powder

Formula: C₁₁H₁₅NO
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00134405
CAS Number: 4411-25-0
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 140 to 150 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR