Dimethylisopropylchlorosilane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorodimethylisopropylsilane, Isopropyldimethylchlorosilane

AC31357-0250
TS31357-0250EA 201 USD
TS31357-0250
Dimethylisopropylchlorosilane 95%
Dimethylisopropylchlorosilane
Formula: C₅H₁₃ClSi
MW: 136.69 g/mol
Boiling Pt: 110…114 °C (760 mmHg)
Density: 0.87 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00009917
CAS Number: 3634-56-8
UN: 2985
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR