(3-Aminopropyl)trimethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane
AC31325-0025
AC31325-1000
AC31325-5000
TS31325-0025EA
765
USD
TS31325-0025
TS31325-1000
TS31325-5000
(3-Aminopropyl)trimethoxysilane 95%
(3-Aminopropyl)trimethoxysilane
Formula:
H₂NCH₂CH₂CH₂Si(OCH₃)₃ MW: 179.29 g/mol Boiling Pt: 194 Density: 1.01 g/cm³ Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008206 CAS Number: 13822-56-5 EINECS: 237-511-5 UN: 3267 ADR: 8 III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4220 to 1.4250 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...