(3-Aminopropyl)trimethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane

AC31325-0025 AC31325-1000 AC31325-5000
TS31325-0025EA 765 USD
TS31325-0025 TS31325-1000 TS31325-5000
(3-Aminopropyl)trimethoxysilane 95%
(3-Aminopropyl)trimethoxysilane
Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
Density: 1.01 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008206
CAS Number: 13822-56-5
EINECS: 237-511-5
UN: 3267
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4250 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR