α,α,α-Trifluoro-m-toluoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride

31294-0050 31294-0250
TS31294-0050EA 30 USD
TS31294-0050 TS31294-0250
α,α,α-Trifluoro-m-toluoyl chloride 98%
α,α,α-Trifluoro-m-toluoyl chloride
Formula: C₈H₄ClF₃O
MW: 208.57
Boiling Pt: 184…186 °C (750 mmHg)
Density: 1.38 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000680
CAS Number: 2251-65-2
EINECS: 218-844-5
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4780 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR