Tetrafluorotetracyanoquinodimethane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, TCNQF4, 2,2'-(Perfluorocyclohexa-2,5-diene-1,4-diylidene)dimalononitrile

31267-0050
TS31267-0050EA 48 USD
TS31267-0050
Tetrafluorotetracyanoquinodimethane 95%
Tetrafluorotetracyanoquinodimethane
Formula: C₁₂F₄N₄
MW: 276.15 g/mol
Melting Pt: 291 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00042382
CAS Number: 29261-33-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow-green to orange-brown Powder
Infrared spectrum Conforms
Assay ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR