5-Fluoroindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Fluoro-1H-indole
31192-0010
31192-0050
31192-0250
31192-2500
TS31192-0010EA
59
USD
TS31192-0010
TS31192-0050
TS31192-0250
TS31192-2500
5-Fluoroindole 98%
5-Fluoroindole
Formula:
C₈H₆FN Melting Pt: 44…48 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005671 CAS Number: 399-52-0 EINECS: 206-917-4 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 44 to 48 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...