2-Ethylpyrrole 98%

Supplier: Thermo Scientific Chemicals
Warning

30877-0010
TS30877-0010EA 100 USD
TS30877-0010
2-Ethylpyrrole 98%
2-Ethylpyrrole
Formula: C₆H₉N
Boiling Pt: 142 °C (15 mmHg)
Density: 0.929 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00060477
CAS Number: 1551-06-0

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4985 to 1.5025 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR