(S)-(+)-β-Aminophenethyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

30757-0010 30757-0050 30757-0250
TS30757-0010EA 34 USD
TS30757-0010 TS30757-0050 TS30757-0250
(S)-(+)-β-Aminophenethyl alcohol 98%
(S)-(+)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS Number: 20989-17-7
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 79 °C
HPLC ≥97.5 %
Specific optical rotation +24.5° to +26.5° (20 °C, 589 nm) (c=6, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR