5-Amino-3-methylisoxazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

305870010
TS305870010EA 29 USD
TS305870010
5-Amino-3-methylisoxazole 98%
5-Amino-3-methylisoxazole
Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 84
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9
UN: 2811

Order Now

Specification Test Results

Appearance Yellow to orange-brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 77 to 84 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR