5-Bromoanthranilic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-5-bromobenzoic acid
30562-0050
30562-0250
TS30562-0050EA
51
USD
TS30562-0050
TS30562-0250
5-Bromoanthranilic acid 97%
5-Bromoanthranilic acid
Formula:
C₇H₆BrNO₂ Melting Pt: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007823 CAS Number: 5794-88-7 EINECS: 227-338-3 Merck Index: 13,01389 |
Specification Test Results
Appearance | Beige Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...