Iron(II) acetate 95%

Supplier: Thermo Scientific Chemicals
Warning

AC30537-0100 AC30537-0500 AC30537-2500
TS30537-0100EA 115 USD
TS30537-0100 TS30537-0500 TS30537-2500
Iron(II) acetate 95%
Iron (II) acetate
Formula: C₄H₆FeO₄
Melting Pt: 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00058909
CAS Number: 3094-87-9
EINECS: 221-441-7

Order Now

Specification Test Results

Appearance Off-white to light gray to light brown Powder
Infrared spectrum Conforms
Titration with KMnO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR