Scopoletin (7-hydroxy-6-methoxycumarin) 95%

Supplier: Thermo Scientific Chemicals
Warning

30290-0010 30290-0500 30290-2500
TS30290-0010EA 762 USD
TS30290-0010 TS30290-0500 TS30290-2500
Scopoletin (7-hydroxy-6-methoxycumarin) 95%
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formula: C₁₀H₈O₄
Melting Pt: 200…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00006872
CAS Number: 92-61-5
EINECS: 202-171-9

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 200 to 207 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR