(±)-α-Terpineol 97+%

Supplier: Thermo Scientific Chemicals
Warning

301610025 301610250 301615000
TS301610025EA 197 USD
TS301610025 TS301610250 TS301615000
(±)-α-Terpineol 97+%
(±)-α-Terpineol
Formula: C₁₀H₁₈O
Boiling Pt: 214…224 °C (760 mmHg)
Melting Pt: 31…34 °C
Density: 0.937 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6

Order Now

Specification Test Results

Appearance Clear colorless to white solid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4850 (20°C, 589 nm)
GC ≥97.0 %
Specific gravity (20°C) 0.934 to 0.940


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR