tert-Butyldichlorophosphine
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
t-Butyldichlorophosphine
30053-0025
30053-0100
TS30053-0025EA
58
USD
TS30053-0025
TS30053-0100
tert-Butyldichlorophosphine
tert-Butyldichlorophosphine
Formula:
C₄H₉Cl₂P Boiling Pt: 142…150 °C (760 mmHg) Melting Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013615 CAS Number: 25979-07-1 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow Low melting crystalline mass |
Infrared spectrum | Conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...