Bis(triphenylphosphine)palladium(II) chloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(triphenylphosphine)palladium(II) dichloride

AC29925-0025 AC29925-0050 AC29925-0250 AC29925-1000
TS29925-0025EA 124 USD
TS29925-0025 TS29925-0050 TS29925-0250 TS29925-1000
Bis(triphenylphosphine)palladium(II) chloride 98%
Bis(triphenylphosphine)palladium (II) chloride
Formula: C₃₆H₃₀Cl₂P₂Pd
Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00009593
CAS Number: 13965-03-2
EINECS: 237-744-2

Order Now

Specification Test Results

Appearance Yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Assay 14.8 to 15.5 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR