trans-2-Nonenal 95%

Supplier: Thermo Scientific Chemicals
Warning

29831-0250
TS29831-0250EA 125 USD
TS29831-0250
trans-2-Nonenal 95%
trans-2-Nonenal
Formula: C₉H₁₆O
Boiling Pt: 98…100 °C (12 mmHg)
Density: 0.846 g/cm³
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007012
CAS Number: 18829-56-6
EINECS: 242-609-6
Merck Index: 12,06771

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4560 (20 °C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR