3-Octylthiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

29614-0100
TS29614-0100EA 142 USD
TS29614-0100
3-Octylthiophene 97%
3-Octylthiophene
Formula: C₁₂H₂₀S
Boiling Pt: 106…107 °C (3 mmHg)
Density: 0.91 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00085281
CAS Number: 65016-62-8
UN: 3334

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4950 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR