2,3-Naphthalenedialdehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

29472-5000 29472-1000
TS29472-5000EA 498 USD
TS29472-5000 TS29472-1000
2,3-Naphthalenedialdehyde 95%
2,3-Naphthalenedialdehyde (NDA)
Formula: C₁₂H₈O₂
Melting Pt: 134…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016618
CAS Number: 7149-49-7

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR