4-Bromo-3-methylanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

294030050 294030250
TS294030050EA 34 USD
TS294030050 TS294030250
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole
Formula: C₈H₉BrO
Boiling Pt: 111…112 °C (15 mmHg)
Density: 0.91 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5620 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR