6-Methylbenzo[d]oxazole 99.5+%

Supplier: Thermo Scientific Chemicals
Warning

29311-0025
TS29311-0025EA 84 USD
TS29311-0025
6-Methylbenzo[d]oxazole 99.5+%
6-Methylbenzo[d]oxazole
Formula: C₈H₇NO
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00216937
CAS Number: 10531-80-3

Order Now

Specification Test Results

Appearance Amber Low melting solid
Infrared spectrum Conforms
Melting point 29 to 31 °C
GC ≥99.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR