4,4-Dimethoxybutanone 92%

Supplier: Thermo Scientific Chemicals
Warning

29233-0010 29233-2500
TS29233-0010EA 296 USD
TS29233-0010 TS29233-2500
4,4-Dimethoxybutanone 92%
4,4-Dimethoxybutanone
Formula: C₆H₁₂O₃
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.99 g/cm³
Flash Pt: 49 °C
MDL Number: MFCD00008789
CAS Number: 5436-21-5
EINECS: 226-605-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4160 (20°C, 589 nm)
GC ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR