Pyridine 99+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

29086-1000 29086-0010 29086-0025 290860100
TS29086-1000EA 45 USD
TS29086-1000 TS29086-0010 TS29086-0025 TS290860100
Pyridine 99+% for analysis
Pyridine
Formula: C₅H₅N
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: –42 °C
Density: 0.978 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
UN: 1282
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤10 APHA
Non-volatile matter ≤0.002 %
Ammonia (NH3) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR