Carbomer 940

Synonyms: Carbopol® 940

27838-2500
TS27838-2500EA 384 USD
TS27838-2500
Carbomer 940
Carbomer 940
Storage Temperature: Ambient
MDL Number: MFCD01941675
CAS Number: 9007-17-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
Heavy metals ≤10 ppm (Hg, Pb, As, Sb)
Viscosity 40000 to 60000 cP (at 25°C) (Brookfield, 20 rpm) (neutralized soln., 0.5% dispersion)
Clarity of solution ≥85 % transmission (neutralized soln., 420 nm, 0.5% dispersion)
Residual solvents ≤0.5 % Benzene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR