7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

27782-2500
TS27782-2500EA 54 USD
TS27782-2500
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 98%
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline
Formula: C₁₀H₁₁NO₂
Melting Pt: 187…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00143517
CAS Number: 4602-73-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder and/or flakes
Infrared spectrum Conforms
Melting point 187 to 191 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR